This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02311015_g1
See other AT5G28830 GE Assays ›
Gene Symbol:
AT5G28830
Gene Name:
calcium-binding EF hand family protein
Gene Aliases:
AT5G28830, F7P1.10, F7P1_10
Chromosome Location:
Chr.5: 10856661 - 10858687 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02311015_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT5G28830
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_122766.4 NP_198235.1 1 - 2 437 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s)

biological_process

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0