This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02299936_g1
See other PGRL1B GE Assays ›
Gene Symbol:
PGRL1B
Gene Name:
PGR5-like B
Gene Aliases:
AT4G11960, F16J13.30, F16J13_30, PGR5-like B
Chromosome Location:
Chr.4: 7174886 - 7177888 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02299936_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGRL1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203773.1 NP_001190702.1 11 - 12 1097 108
NM_117266.4 NP_192933.2 10 - 11 1134 108
GenBank mRNA AK117721.1 - 10 - 11 1004 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

photosynthetic electron transport in photosystem I
photosynthesis
oxidoreductase activity, acting on iron-sulfur proteins as donors

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0