This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02299901_g1
See other CCS52A2 GE Assays ›
Gene Symbol:
CCS52A2
Gene Name:
cell cycle switch protein 52 A2
Gene Aliases:
AT4G11920, FIZZY-RELATED 1, FZR1, T26M18.130, T26M18_130, cell cycle switch protein 52 A2, cell cycle switch protein 52 A2
Chromosome Location:
Chr.4: 7160243 - 7163427 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02299901_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCS52A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_117262.3 NP_192929.2 1 - 2 646 92
GenBank mRNA AY063875.1 - 1 - 2 601 92
AY091235.1 - 1 - 2 476 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell size
maintenance of shoot apical meristem identity
protein ubiquitination
anaphase-promoting complex-dependent catabolic process
positive regulation of DNA endoreduplication
positive regulation of ubiquitin protein ligase activity
positive regulation of anaphase-promoting complex-dependent catabolic process
protein binding
anaphase-promoting complex binding
ubiquitin ligase activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0