This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02290048_g1
See other CMT1 GE Assays ›
Gene Symbol:
CMT1
Gene Name:
chromomethylase 1
Gene Aliases:
AT1G80740, CHROMOMETHYLASE 1, DMT4, DNA METHYLTRANSFERASE 4, F23A5.9, F23A5_9, chromomethylase 1
Chromosome Location:
Chr.1: 30342394 - 30347048 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02290048_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CMT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_106722.2 NP_565245.1 11 - 12 1545 156

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein DNA methyltransferase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA mediated transformation
chromatin binding
DNA (cytosine-5-)-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0