This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02287761_s1
See other AAE2 GE Assays ›
Gene Symbol:
AAE2 AT1G77230
Gene Name:
AMP-dependent synthetase and ligase family protein
Tetratricopeptide repeat (TPR)-like superfamily protein
Gene Aliases:
AT1G77240, T14N5.10, T14N5_10, acyl activating enzyme 2
AT1G77230, T14N5.9, T14N5_9
Chromosome Location:
Chr.1: 29017958 - 29019789 on Build TAIR10.1
Chromosome Location:
Chr.1: 29016120 - 29019768 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
-
Assay ID At02287761_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AAE2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_106373.2 NP_177848.1 1 1647 85
Gene Symbol: AT1G77230
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_106372.2 NP_177847.2 5 936 85
GenBank mRNA AK175648.1 - 5 935 85
AK175665.1 - 5 935 85
AY064038.1 - 5 935 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ligase

Gene Ontology Categories:

Function(s)

biological_process

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0