This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02284163_s1
See other RPS5 GE Assays ›
Gene Symbol:
RPS5
Gene Name:
Disease resistance protein (CC-NBS-LRR class) family
Gene Aliases:
AT1G12220, DISEASE RESISTANCE PROTEIN RPS5, RESISTANT TO P. SYRINGAE 5, T28K15.5, T28K15_5
Chromosome Location:
Chr.1: 4144257 - 4147939 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02284163_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001198041.2 NP_001184970.1 2 3000 110
NM_001332024.1 NP_001323155.1 2 2571 110
NM_101094.3 NP_172686.1 1 2844 110
GenBank mRNA AK221293.1 - 1 2697 110
BT010583.1 - 1 2621 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

antimicrobial response protein defense/immunity protein

Gene Ontology Categories:

Function(s) Process(es)

defense response
cell death
plant-type hypersensitive response
defense response to bacterium, incompatible interaction
nucleotide binding
protein binding
signaling receptor activity
identical protein binding
ADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0