This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02284082_g1
See other FMO GS-OX5 GE Assays ›
Gene Symbol:
FMO GS-OX5
Gene Name:
flavin-monooxygenase glucosinolate S-oxygenase 5
Gene Aliases:
AT1G12140, T28K15.12, T28K15_12, flavin-monooxygenase glucosinolate S-oxygenase 5
Chromosome Location:
Chr.1: 4121285 - 4124595 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02284082_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FMO GS-OX5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001084053.1 NP_001077522.1 4 - 5 999 74
NM_001332018.1 NP_001323223.1 4 - 5 1005 74
NM_101086.5 NP_172678.3 4 - 5 1089 74
GenBank mRNA BT030387.1 - 4 - 5 988 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

glucosinolate biosynthetic process
regulation of organ growth
regulation of hormone biosynthetic process
floral organ morphogenesis
oxidation-reduction process
monooxygenase activity
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding
8-methylthiopropyl glucosinolate S-oxygenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0