This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02278165_s1
See other AT3G22670 GE Assays ›
Gene Symbol:
AT3G22670 RDM1
Gene Name:
Pentatricopeptide repeat (PPR) superfamily protein
RNA-DIRECTED DNA METHYLATION 1
Gene Aliases:
AT3G22670
AT3G22680, RNA-DIRECTED DNA METHYLATION 1
Chromosome Location:
Chr.3: 8017654 - 8019552 on Build TAIR10.1
Chromosome Location:
Chr.3: 8017633 - 8020508 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
-
Assay ID At02278165_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT3G22670
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_113166.2 NP_188906.1 1 1721 103
Gene Symbol: RDM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_113167.5 NP_188907.2 1 200 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_113167.5 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

biological_process
DNA methylation
regulation of DNA methylation
production of small RNA involved in gene silencing by RNA
protein binding
protein self-association

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header