This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02273350_g1
See other ASP3 GE Assays ›
Gene Symbol:
ASP3
Gene Name:
aspartate aminotransferase 3
Gene Aliases:
ASPARTATE AMINOTRANSFERASE, AT5G11520, CHLOROPLAST PRECURSOR, F15N18.110, F15N18_110, YELLOW-LEAF-SPECIFIC GENE 4, YLS4, aspartate aminotransferase 3
Chromosome Location:
Chr.5: 3684946 - 3687854 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02273350_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_121190.3 NP_196713.1 1 - 2 322 86
GenBank mRNA AY050765.1 - 1 - 2 232 86
AY079310.1 - 1 - 2 189 86
U15034.1 - 1 - 2 232 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

2-oxoglutarate metabolic process
aspartate metabolic process
glutamate metabolic process
biosynthetic process
leaf senescence
L-aspartate:2-oxoglutarate aminotransferase activity
pyridoxal phosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0