This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02264986_s1
See other ADT6 GE Assays ›
Gene Symbol:
ADT6 TIL1
Gene Name:
arogenate dehydratase 6
DNA polymerase epsilon catalytic subunit
Gene Aliases:
AT1G08250, Arabidopsis thaliana arogenate dehydratase 6, AtADT6, T23G18.10, T23G18_10, arogenate dehydratase 6
ABA OVERLY SENSITIVE 4, ABO4, AT1G08260, EARLY IN SHORT DAYS 7, EMB142, EMB2284, EMB529, EMBRYO DEFECTIVE 142, EMBRYO DEFECTIVE 2284, EMBRYO DEFECTIVE 529, ESD7, POL2A, T23G18.21, T23G18_21, TILTED 1
Chromosome Location:
Chr.1: 2588776 - 2590609 on Build TAIR10.1
Chromosome Location:
Chr.1: 2588854 - 2606892 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
-
Assay ID At02264986_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADT6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_100698.4 NP_563809.1 1 1656 120
Gene Symbol: TIL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001331765.1 NP_001322767.1 1 101 120
NM_100699.6 NP_172303.5 1 101 120

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001331765.1, NM_100699.6 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

dehydratase DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

L-phenylalanine biosynthetic process
mitotic cell cycle
leading strand elongation
base-excision repair, gap-filling
nucleotide-excision repair, DNA gap filling
embryo development ending in seed dormancy
embryonic root morphogenesis
DNA replication proofreading
negative regulation of long-day photoperiodism, flowering
regulation of cell division
prephenate dehydratase activity
arogenate dehydratase activity
nucleotide binding
DNA binding
DNA-directed DNA polymerase activity
protein binding
zinc ion binding
single-stranded DNA 3'-5' exodeoxyribonuclease activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header