This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02239702_g1
See other TPS4 GE Assays ›
Gene Symbol:
TPS4
Gene Name:
trehalose-6-phosphatase synthase S4
Gene Aliases:
ARABIDOPSIS THALIANA TREHALOSE PHOSPHATASE/SYNTHASE 4, AT4G27550, ATTPS4, T29A15.40, T29A15_40, trehalose-6-phosphatase synthase S4
Chromosome Location:
Chr.4: 13755649 - 13759740 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02239702_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TPS4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_118890.2 NP_194485.1 10 - 11 1433 104
GenBank mRNA KJ676985.1 - 10 - 11 1393 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

trehalose biosynthetic process
trehalose metabolism in response to stress
alpha,alpha-trehalose-phosphate synthase (UDP-forming) activity
transferase activity, transferring glycosyl groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0