This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02236624_s1
See other RPM1 GE Assays ›
Gene Symbol:
RPM1
Gene Name:
NB-ARC domain-containing disease resistance protein
Gene Aliases:
AT3G07040, DISEASE RESISTANCE PROTEIN RPM1, RESISTANCE TO P. SYRINGAE PV MACULICOLA 1, RESISTANCE TO PSEUDOMONAS SYRINGAE 3, RPS3
Chromosome Location:
Chr.3: 2225855 - 2229555 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02236624_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_111584.3 NP_187360.1 1 3494 140

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

antimicrobial response protein defense/immunity protein

Gene Ontology Categories:

Function(s) Process(es)

defense response
plant-type hypersensitive response
nucleotide binding
protein binding
ADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0