This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02228192_s1
See other AT1G72290 GE Assays ›
Gene Symbol:
AT1G72290
Gene Name:
Kunitz family trypsin and protease inhibitor protein
Gene Aliases:
AT1G72290, T9N14.19, T9N14_19
Chromosome Location:
Chr.1: 27215787 - 27216857 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02228192_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT1G72290
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_105888.2 NP_177373.1 1 957 112
GenBank mRNA AY084376.1 - 1 956 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

response to wounding
response to insect
negative regulation of peptidase activity
regulation of cell death
response to herbivore
regulation of pollen tube growth
cysteine-type endopeptidase inhibitor activity
protein binding
peptidase inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0