This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02224285_g1
See other FMO GS-OX1 GE Assays ›
Gene Symbol:
FMO GS-OX1
Gene Name:
flavin-monooxygenase glucosinolate S-oxygenase 1
Gene Aliases:
AT1G65860, F12P19.2, F12P19_2, flavin-monooxygenase glucosinolate S-oxygenase 1
Chromosome Location:
Chr.1: 24498954 - 24502776 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02224285_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FMO GS-OX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_105258.3 NP_176761.1 5 - 6 1089 78
GenBank mRNA BT000473.1 - 5 - 6 1059 78
BT002190.1 - 5 - 6 991 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

glucosinolate biosynthetic process from homomethionine
oxidation-reduction process
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding
3-methylthiopropyl glucosinolate S-oxygenase activity
4-methylthiopropyl glucosinolate S-oxygenase activity
5-methylthiopropyl glucosinolate S-oxygenase activity
6-methylthiopropyl glucosinolate S-oxygenase activity
7-methylthiopropyl glucosinolate S-oxygenase activity
8-methylthiopropyl glucosinolate S-oxygenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0