This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02220605_s1
See other HCD1 GE Assays ›
Gene Symbol:
HCD1
Gene Name:
3-hydroxyacyl-CoA dehydratase 1
Gene Aliases:
3-hydroxyacyl-CoA dehydratase 1, ARABIDOPSIS THALIANA DELTA(3), AT4G14440, ATECI3, DELTA(2)-ENOYL COA ISOMERASE 3, DELTA(3), DL3260C, ECI3, FCAALL.212
Chromosome Location:
Chr.4: 8306590 - 8307934 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02220605_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HCD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_117523.4 NP_193180.1 1 1039 99
GenBank mRNA AY072398.1 - 1 858 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

hydratase

Gene Ontology Categories:

Function(s) Process(es)

fatty acid beta-oxidation
fatty acid catabolic process
response to karrikin
dodecenoyl-CoA delta-isomerase activity
enoyl-CoA hydratase activity
carnitine racemase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0