This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02219336_s1
See other ORC1B GE Assays ›
Gene Symbol:
ORC1B
Gene Name:
origin of replication complex 1B
Gene Aliases:
AT4G12620, ATORC1B, T1P17.210, T1P17_210, UNE13, UNFERTILIZED EMBRYO SAC 13, origin of replication complex 1B
Chromosome Location:
Chr.4: 7459609 - 7462447 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02219336_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ORC1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_117332.3 NP_192999.1 1 531 139
GenBank mRNA AJ426477.1 - 1 337 139

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

replication origin binding protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic cell cycle
DNA replication
DNA replication initiation
regulation of transcription, DNA-templated
double fertilization forming a zygote and endosperm
mitotic DNA replication checkpoint
chromatin binding
DNA replication origin binding
protein binding
double-stranded methylated DNA binding
histone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0