This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02218251_g1
See other NPR1 GE Assays ›
Gene Symbol:
NPR1
Gene Name:
regulatory protein (NPR1)
Gene Aliases:
ARABIDOPSIS NONEXPRESSER OF PR GENES 1, AT1G64280, ATNPR1, F15H21.6, F15H21_6, NIM1, NON-INDUCIBLE IMMUNITY 1, NONEXPRESSER OF PR GENES 1, REGULATORY PROTEIN NPR1, SAI1, SALICYLIC ACID INSENSITIVE 1
Chromosome Location:
Chr.1: 23852748 - 23855658 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02218251_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NPR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_105102.3 NP_176610.1 2 - 3 1547 75
GenBank mRNA AY050455.1 - 2 - 3 1454 75
AY093992.1 - 2 - 3 1296 75
HQ845963.1 - 2 - 3 1296 75
U76707.1 - 2 - 3 1388 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
cell death
response to heat
response to wounding
response to bacterium
response to insect
systemic acquired resistance
induced systemic resistance
systemic acquired resistance, salicylic acid mediated signaling pathway
regulation of systemic acquired resistance
protein ubiquitination
negative regulation of defense response
defense response to bacterium
positive regulation of transcription, DNA-templated
defense response to fungus
regulation of jasmonic acid mediated signaling pathway
regulation of salicylic acid mediated signaling pathway
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0