This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02216215_s1
See other AT1G62500 GE Assays ›
Gene Symbol:
AT1G62500
Gene Name:
Bifunctional inhibitor/lipid-transfer protein/seed storage 2S albumin superfamily protein
Gene Aliases:
AT1G62500, T3P18.6, T3P18_6
Chromosome Location:
Chr.1: 23131928 - 23133507 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02216215_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT1G62500
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_104929.3 NP_176439.1 1 1504 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0