This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02213005_g1
See other TBP2 GE Assays ›
Gene Symbol:
TBP2
Gene Name:
TATA binding protein 2
Gene Aliases:
A. THALIANA TATA BINDING PROTEIN 2, AT1G55520, ATTBP2, T5A14.8, T5A14_8, TATA binding protein 2
Chromosome Location:
Chr.1: 20725470 - 20728093 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02213005_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TBP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001333716.1 NP_001321102.1 6 - 7 598 115
NM_001333717.1 NP_001321103.1 6 - 7 600 115
NM_104427.4 NP_175948.1 6 - 7 598 115
NM_179481.2 NP_849812.1 6 - 7 598 115
GenBank mRNA AB493506.1 - 5 - 6 331 115
AK227470.1 - 6 - 7 617 115
AY463626.1 - 5 - 6 331 115
BT006488.1 - 5 - 6 331 115
X54995.1 - 585 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

DNA-templated transcription, initiation
DNA binding
protein binding
obsolete general RNA polymerase II transcription factor activity
TBP-class protein binding
general transcription initiation factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0