This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02203352_g1
See other FRS2 GE Assays ›
Gene Symbol:
FRS2
Gene Name:
FAR1-related sequence 2
Gene Aliases:
AT2G32250, FAR1-related sequence 2, T32F6.23, T32F6_23
Chromosome Location:
Chr.2: 13693094 - 13697206 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02203352_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FRS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001084522.2 NP_001077991.1 5 - 6 2518 76
NM_001124959.2 NP_001118431.1 4 - 5 2640 76
NM_001336379.1 NP_001324563.1 4 - 5 2629 76
NM_001336380.1 NP_001324564.1 4 - 5 2629 76
NM_001336381.1 NP_001324561.1 5 - 6 2315 76
NM_001336382.1 NP_001324562.1 5 - 6 2315 76
NM_128784.4 NP_180784.2 4 - 5 2629 76
NM_201851.2 NP_973580.1 4 - 5 2629 76
GenBank mRNA AK228730.1 - 4 - 5 2638 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0