This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02201505_g1
See other TRP1 GE Assays ›
Gene Symbol:
TRP1
Gene Name:
tryptophan biosynthesis 1
Gene Aliases:
AT5G17990, MCM23.6, MCM23_6, PHOSPHORIBOSYLANTHRANILATE TRANSFERASE, PHOSPHORIBOSYLANTHRANILATE TRANSFERASE 1, pat1, tryptophan biosynthesis 1
Chromosome Location:
Chr.5: 5957080 - 5960201 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02201505_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_121804.5 NP_197300.1 6 - 7 1245 106
GenBank mRNA AY054506.1 - 6 - 7 1109 106
AY093298.1 - 6 - 7 995 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

tryptophan biosynthetic process
anthranilate phosphoribosyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0