This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02201414_g1
See other DAR4 GE Assays ›
Gene Symbol:
DAR4
Gene Name:
DA1-related protein 4
Gene Aliases:
AT5G17890, CHILLING SENSITIVE 3, CHS3, DA1-related protein 4, MPI7.6, MPI7_6
Chromosome Location:
Chr.5: 5916862 - 5923494 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02201414_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DAR4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_121795.4 NP_197291.2 10 - 11 4475 160

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

defense response
response to cold
ubiquitin binding
ADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0