This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02174736_g1
See other HCC2 GE Assays ›
Gene Symbol:
HCC2
Gene Name:
Thioredoxin superfamily protein
Gene Aliases:
AT4G39740, T19P19.130, T19P19_130, homologue of copper chaperone SCO1 2
Chromosome Location:
Chr.4: 18435293 - 18437258 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02174736_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HCC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_120135.5 NP_568068.1 2 - 3 371 60
GenBank mRNA AY087747.1 - 2 - 3 238 60
BT024500.1 - 2 - 3 208 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial respiratory chain complex IV assembly
copper ion homeostasis
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0