Assay Details
Gene Symbol: TRFL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148439.4 | NP_683280.1 | 4 - 5 | 1219 | 96 | |
GenBank mRNA | AY519534.1 | - | 4 - 5 | 1111 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
