This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02165743_s1
See other AT4G22470 GE Assays ›
Gene Symbol:
AT4G22470 AT4G22467
Gene Name:
protease inhibitor/seed storage/lipid transfer protein (LTP) family protein
hypothetical protein
Gene Aliases:
AT4G22470, F7K2.50, F7K2_50
AT4G22467
Chromosome Location:
Chr.4: 11840134 - 11841450 on Build TAIR10.1
Chromosome Location:
Chr.4: 11840119 - 11841517 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
-
Assay ID At02165743_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT4G22470
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_118373.3 NP_193978.2 1 1268 162
Gene Symbol: AT4G22467
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001341545.1 NP_001329811.1 1 65 162

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001341545.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of systemic acquired resistance
biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0