This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02163813_m1
See other NPC3 GE Assays ›
Gene Symbol:
NPC3
Gene Name:
non-specific phospholipase C3
Gene Aliases:
AT3G03520, T21P5.6, T21P5_6, non-specific phospholipase C3
Chromosome Location:
Chr.3: 837805 - 840645 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02163813_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NPC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001337474.1 NP_001326666.1 2 - 3 758 126
NM_111223.5 NP_187002.1 2 - 3 775 126
GenBank mRNA AB084295.1 - 2 - 3 641 126
AY150477.1 - 2 - 3 641 126

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

phosphate-containing compound metabolic process
phospholipid catabolic process
cellular response to phosphate starvation
phospholipase C activity
hydrolase activity, acting on ester bonds
lysophosphatidic acid phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0