This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02163024_g1
See other BIG GE Assays ›
Gene Symbol:
BIG
Gene Name:
auxin transport protein (BIG)
Gene Aliases:
ASA1, AT3G02260, ATTENUATED SHADE AVOIDANCE 1, CORYMBOSA1, CRM1, DARK OVER-EXPRESSION OF CAB 1, DOC1, F14P3.3, F14P3_3, LOW PHOSPHATE-RESISTANT ROOT 1, LPR1, TIR3, TRANSPORT INHIBITOR RESPONSE 3, UMB1, UMBRELLA 1
Chromosome Location:
Chr.3: 430877 - 448680 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02163024_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BIG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001337388.1 NP_001327521.1 11 - 12 13095 115
NM_001337389.1 NP_001327522.1 11 - 12 13023 115
NM_001337390.1 NP_001327520.1 11 - 12 12966 115
NM_111093.2 NP_186875.2 11 - 12 12967 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

response to fungus
photomorphogenesis
response to auxin
unidimensional cell growth
auxin polar transport
lateral root formation
inflorescence morphogenesis
indeterminate inflorescence morphogenesis
root development
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0