This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02162849_s1
See other AT3G02010 GE Assays ›
Gene Symbol:
AT3G02010 AK3
Gene Name:
Pentatricopeptide repeat (PPR) superfamily protein
aspartate kinase 3
Gene Aliases:
AT3G02010, F1C9.21
AT3G02020, F1C9.20, F1C9_20, aspartate kinase 3
Chromosome Location:
Chr.3: 337906 - 340442 on Build TAIR10.1
Chromosome Location:
Chr.3: 337712 - 343912 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
-
Assay ID At02162849_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT3G02010
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_111067.2 NP_186850.1 1 2408 117
Gene Symbol: AK3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_111068.4 NP_186851.1 13 2505 117

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

amino acid kinase

Gene Ontology Categories:

Function(s) Process(es)

RNA modification
aspartate family amino acid biosynthetic process
threonine biosynthetic process
lysine biosynthetic process via diaminopimelate
homoserine biosynthetic process
zinc ion binding
aspartate kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0