This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02156629_g1
See other ABA3 GE Assays ›
Gene Symbol:
ABA3
Gene Name:
molybdenum cofactor sulfurase (LOS5) (ABA3)
Gene Aliases:
ABA DEFICIENT 3, ACI2, ALTERED CHLOROPLAST IMPORT 2, AT1G16540, ATABA3, AtLOS5, F19K19.13, F19K19_13, LOS5, LOW OSMOTIC STRESS 5, MOLYBDENUM COFACTOR SULFURASE, SIR3, SIRTINOL RESISTANT 3
Chromosome Location:
Chr.1: 5659370 - 5665488 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02156629_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABA3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001332230.1 NP_001322901.1 7 - 8 959 89
NM_001332231.1 NP_001322902.1 7 - 8 959 89
NM_101519.3 NP_564001.1 7 - 8 959 89
GenBank mRNA AF325457.1 - 7 - 8 914 89
AY034895.1 - 7 - 8 864 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to osmotic stress
response to heat
response to cold
response to salt stress
abscisic acid biosynthetic process
auxin-activated signaling pathway
response to glucose
stomatal movement
sugar mediated signaling pathway
molybdenum incorporation into molybdenum-molybdopterin complex
defense response to bacterium
protein import into chloroplast stroma
molecular_function
Mo-molybdopterin cofactor sulfurase activity
selenocysteine lyase activity
molybdenum cofactor sulfurtransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0