Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Proteins & Peptides ›
  • THAP5 Proteins

Invitrogen

Human THAP5 (aa 88-159) Control Fragment Recombinant Protein

View all (2) THAP5 proteins

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Tech Support
Datasheet
Tech Support

Cite Human THAP5 (aa 88-159) Control Fragment Recombinant Protein

Product Details

RP-104923

Applications
Tested Dilution
Publications

Control (Ctrl)

Assay-dependent
-

Blocking Assay (BLOCK)

Assay-dependent
-
Product Specifications

Species

Human

Expression System

E. coli

Amino acid sequence

EDNQGKDPSKKKSQKKNLEDEKEVCPKAKSEESFVLNETKKNIVNTDVPHQHPELLHSSSLVKPPAPKTGSI

Tag

His-ABP-tag

Class

Recombinant

Type

Protein

Purity

>80% by SDS-PAGE and Coomassie blue staining

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

≥5.0 mg/mL

Purification

purified

Storage buffer

PBS/1M urea, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Product Specific Information

Highest antigen sequence indentity to the following orthologs: Mouse (28%), Rat (28%).

This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65661. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Target Information

Has sequence-specific DNA-binding activity and can function as transcriptional repressor (in vitro). May be a regulator of cell cycle: THAP5 overexpression in human cell lines causes cell cycle arrest at G2/M phase.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: THAP domain-containing protein 5

View more View less

Gene Aliases: THAP5

View more View less

UniProt ID: (Human) Q7Z6K1

View more View less

Entrez Gene ID: (Human) 168451

View more View less

It has to be done as per old AB suggested Products section.
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-569546dcc-wtrnc:80/100.66.131.99:80.
git-commit: 347fb3dda4b74a768f64101cbb40f39f590272fd
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0