Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Proteins & Peptides ›
  • Paxillin Proteins

Invitrogen

Human Paxillin Control Fragment Recombinant Protein

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Tech Support
Datasheet
Tech Support

Cite Human Paxillin Control Fragment Recombinant Protein

Product Details

RP-104919

Applications
Tested Dilution
Publications

Control (Ctrl)

Assay-dependent
-

Blocking Assay (BLOCK)

Assay-dependent
-
Product Specifications

Species

Human

Expression System

E. coli

Amino acid sequence

ALNGTILDPLDQWQPSGSRFIHQQPQSSSPVYGSSAKTSSVSNPQDSVGSPCSRVGEEEHVYSFPNKQKSAEPSPTVMSTSL

Tag

His-ABP-tag

Class

Recombinant

Type

Protein

Purity

>80% by SDS-PAGE and Coomassie blue staining

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

≥5.0 mg/mL

Purification

purified

Storage buffer

1M urea/PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Product Specific Information

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%).

This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111334. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Target Information

Paxillin is a focal adhesion protein and a substrate for several tyrosine kinases such as src, FAK, and p210BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. Paxillin associates tightly with FAK and Crk through its SH2 domain. This interaction is independent of the extracellular matrix. Although paxillin was initially discovered in fibroblasts, its phosphorylation may also be important during neurite extension during differentiation.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: FLJ16691; hCG_1778014; PAX-1; PAXI; Paxillin; paxillin alpha; testicular tissue protein Li 134

View more View less

Gene Aliases: PXN

View more View less

UniProt ID: (Human) P49023

View more View less

Entrez Gene ID: (Human) 5829

View more View less

It has to be done as per old AB suggested Products section.
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm