Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Proteins & Peptides ›
  • PIK3CD/PIK3R1 Proteins

Invitrogen

Human PIK3CD/PIK3R1 (p110 delta/p85 alpha), His Tag Recombinant Protein

View all (2) PIK3CD/PIK3R1 proteins

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Tech Support
Datasheet
Tech Support

Cite Human PIK3CD/PIK3R1 (p110 delta/p85 alpha), His Tag Recombinant Protein

Product Details

PV6452

Applications
Tested Dilution
Publications

Kinase Assay (KA)

Assay-dependent
-
Product Specifications

Species

Human

Expression System

Insect cells

Amino acid sequence

Full length

Tag

His-tag

Molecular weight

84.7-120.6 kDa

Class

Recombinant

Type

Protein

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

purified

Storage buffer

50mM sodium phosphate, pH 7, with 150mM Imidazole, 25% glycerol, 0.25mM DTT, 0.1mM PMSF, 300mM NaCl

Contains

no preservative

Storage conditions

-80°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Dry ice

Product Specific Information

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 µg, 10 µg and 20 µg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 µL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase.

PIK3CD/PIK3R1 (p110-delta/p85-alpha) recombinant human protein is:
• Available in 3 convenient sizes
• Validated in our Adapta™ and Kinase Binding assays
• A full length protein that can help advance your research when used with validated assays

Related assay platforms:
LanthaScreen™ Kinase Binding Assay
Adapta™ Kinase Activity Assay

Protein Form: Full Length

The gene accession number for PIK3CD (p110δ) is NP_005017.3 and for PIK3R1 (p85α) is NP_852664.1.

Target Information

Phosphatidylinositol 3-kinase (PI3K) is composed of a catalytic subunit (p110-delta) and a regulatory subunit (p85-alpha in this case), which are encoded by the PIK3CD and PIK3R1 genes. PI3K phosphorylates phosphoinositides on the 3-hydroxyl group of the inositol ring. It plays a role in oncogenic transformation and is consistently expressed at a high level in AML blast cells. PI3K can play a selective role in the amplification of phosphatidylinositol (3,4,5) trisphosphate (PIP3) levels that leads to neutrophil polarization, and regulates directional migration, but not random locomotion. The PI3K signaling pathway is involved in immune responses relevant to the pathogenesis of rheumatoid arthritis and other inflammatory diseases.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha; Phosphatidylinositol 3-kinase regulatory subunit alpha; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); phosphatidylinositol 3-kinase-associated p-85 alpha; Phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit delta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit delta; phosphoinositide-3-kinase C; phosphoinositide-3-kinase regulatory subunit; phosphoinositide-3-kinase regulatory subunit alpha; phosphoinositide-3-kinase, catalytic, delta polypeptide variant p37delta; phosphoinositide-3-kinase, regulatory subunit 1 (alpha); PI3-kinase p110 subunit delta; PI3-kinase regulatory subunit alpha; PI3-kinase subunit delta; PI3-kinase subunit p85-alpha; PI3K regulatory subunit alpha; PI3Kdelta; PIK3CD; PIK3R1; ptdIns-3-kinase regulatory subunit alpha; PtdIns-3-kinase subunit p110-delta

View more View less

Gene Aliases: AGM7; APDS; GRB1; IMD14; IMD36; p110D; P110DELTA; p85; p85-ALPHA; PI3K; PIK3CD; PIK3R1

View more View less

UniProt ID: (Human) O00329, (Human) P27986

View more View less

Entrez Gene ID: (Human) 5293, (Human) 5295

View more View less

It has to be done as per old AB suggested Products section.
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0