Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Proteins & Peptides ›
  • PHLDA1 Proteins

Invitrogen

Human PHLDA1 (aa 246-307) Control Fragment Recombinant Protein

View all (2) PHLDA1 proteins

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Tech Support
Datasheet
Tech Support

Cite Human PHLDA1 (aa 246-307) Control Fragment Recombinant Protein

Product Details

RP-107243

Applications
Tested Dilution
Publications

Control (Ctrl)

Assay-dependent
-

Blocking Assay (BLOCK)

Assay-dependent
-
Product Specifications

Species

Human

Expression System

E. coli

Amino acid sequence

KGKYMYFTVVMAEGKEIDFRCPQDQGWNAEITLQMVQYKNRQAILAVKSTRQKQQHLVQQQP

Tag

His-ABP-tag

Class

Recombinant

Type

Protein

Purity

>80% by SDS-PAGE and Coomassie blue staining

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

≥5.0 mg/mL

Purification

purified

Storage buffer

PBS/1M urea, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Product Specific Information

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%).

This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84598. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Target Information

Cytotoxic T lymphocyte (CTL)-mediated cytotoxicity constitutes an important component of specific effector mechanisms in immunosurveillance against virus-infected or -transformed cells. Two mechanisms appear to account for this activity, one of which is the perforin-based process. Independently, a FAS-based mechanism involves the transducing molecule FAS (APO-1) and its ligand (FAS-L). The human FAS (APO-1) protein is a 48 kDa cell surface glycoprotein that belongs to a family of receptors that includes CD40, nerve growth factor receptors and tumor necrosis factor receptors. The FAS antigen is expressed on a broad range of lymphoid cell lines, and is expressed at high levels in T cells subsequent to crosslinking of the T cell receptor (TCR). A previously undescribed protein, TDAG51, restores activation- induced apoptosis in cells that have lost the ability to display Fas in response to activation.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Apoptosis-associated nuclear protein; Pleckstrin homology-like domain family A member 1; pleckstrin homology-like domain, family A, member 1; PQ-rich protein; PQR protein; Proline- and glutamine-rich protein; Proline- and histidine-rich protein; proline-histidine rich protein; T-cell death-associated gene 51 protein

View more View less

Gene Aliases: DT1P1B11; PHLDA1; PHRIP; TDAG51

View more View less

UniProt ID: (Human) Q8WV24

View more View less

Entrez Gene ID: (Human) 22822

View more View less

It has to be done as per old AB suggested Products section.
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0