Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Proteins & Peptides ›
  • MARK1 Proteins

Invitrogen

Human MARK1 (MARK), GST Tag Recombinant Protein

View all (3) MARK1 proteins

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Tech Support
Datasheet
Tech Support

Cite Human MARK1 (MARK), GST Tag Recombinant Protein

Product Details

PV4395

Applications
Tested Dilution
Publications

Kinase Assay (KA)

Assay-dependent
-
Product Specifications

Species

Human

Expression System

Baculovirus

Amino acid sequence

Full length

Tag

GST-tag

Molecular weight

116.5 kDa

Class

Recombinant

Type

Protein

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

purified

Storage buffer

50mM tris, pH 7.5, with 0.5mM EDTA, 150mM NaCl, 50% glycerol, 0.02% Triton X-100, 2mM DTT

Contains

no preservative

Storage conditions

-80°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Dry ice

Product Specific Information

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 µg, 10 µg and 20 µg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 µL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase.

Protein Form: Full Length

Target Information

MARK1 (MARK) is a serine/threonine kinase which is involved in cytoskeletal organization and biogenesis. Involved in cell polarity by phosphorylating the microtubule-associated proteins MAP2, MAP4 and MAPT/TAU at KXGS motifs, causing detachment from microtubules, and their disassembly. MARK1 is involved in the regulation of neuronal migration through its dual activities in regulating cellular polarity and microtubule dynamics, possibly by phosphorylating and regulating DCX. Also acts as a positive regulator of the Wnt signaling pathway, probably by mediating phosphorylation of dishevelled proteins (DVL1, DVL2 and/or DVL3).

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: MAP/microtubule affinity-regulating kinase 1; MARK1; Par-1c; PAR1 homolog c; Serine/threonine-protein kinase MARK1

View more View less

Gene Aliases: KIAA1477; MARK; MARK1; Par-1c; Par1c

View more View less

UniProt ID: (Human) Q9P0L2

View more View less

Entrez Gene ID: (Human) 4139

View more View less

It has to be done as per old AB suggested Products section.
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0