Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Proteins & Peptides ›
  • CTRL Proteins

Invitrogen

Human CTRL (aa 128-212) Control Fragment Recombinant Protein

View all (2) CTRL proteins

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Tech Support
Datasheet
Tech Support

Cite Human CTRL (aa 128-212) Control Fragment Recombinant Protein

Product Details

RP-95843

Applications
Tested Dilution
Publications

Control (Ctrl)

Assay-dependent
-

Blocking Assay (BLOCK)

Assay-dependent
-
Product Specifications

Species

Human

Expression System

E. coli

Amino acid sequence

ASPAQYTTRISPVCLASSNEALTEGLTCVTTGWGRLSGVGNVTPAHLQQVALPLVTVNQCRQYWGSSITDSMICAGGAGASSCQG

Tag

His-ABP-tag

Class

Recombinant

Type

Protein

Purity

>80% by SDS-PAGE and Coomassie blue staining

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

≥5.0 mg/mL

Purification

purified

Storage buffer

1M urea/PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Product Specific Information

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%).

This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56959. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Target Information

The CTRL gene encodes chymotrypsin-like protein, a serine protease involved in the digestion of proteins by cleaving peptide bonds. It belongs to the chymotrypsin family of enzymes that are predominantly expressed in the pancreas and play crucial roles in the breakdown of dietary proteins in the small intestine. CTRL functions by hydrolyzing specific peptide bonds, contributing to efficient protein digestion and nutrient absorption. This enzyme's activity is tightly regulated to prevent premature activation within the pancreas, which could lead to pancreatitis or other digestive disorders. The study of the CTRL gene is significant in understanding the mechanisms of digestive health and in diagnosing pancreatic disorders. Furthermore, CTRL's enzyme activity and its role in proteolytic pathways make it an important target for research in digestive enzyme supplementation therapies and treatment strategies for conditions involving impaired protein digestion.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: chymotrypsin-like; Chymotrypsin-like protease CTRL-1

View more View less

Gene Aliases: CTRL; CTRL1

View more View less

UniProt ID: (Human) P40313

View more View less

Entrez Gene ID: (Human) 1506

View more View less

It has to be done as per old AB suggested Products section.
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5cc698dfdd-6b6sp:80/100.66.129.221:80.
git-commit: 7dd0da31084df89e35c5ffd4006b5f1c529b0b63
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0