Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Proteins & Peptides ›
  • CDK11A/CDK11B Proteins

Invitrogen

Human CDK11 (Inactive), GST Tag Recombinant Protein

View all (2) CDK11A/CDK11B proteins

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Tech Support
Datasheet
Tech Support

Cite Human CDK11 (Inactive), GST Tag Recombinant Protein

Product Details

PV6283

Applications
Tested Dilution
Publications

Kinase Assay (KA)

Assay-dependent
-
Product Specifications

Species

Human

Expression System

Baculovirus

Amino acid sequence

1-795

Tag

GST-tag

Molecular weight

84.2 kDa

Class

Recombinant

Type

Protein

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

purified

Storage buffer

50mM tris HCl, pH 7.5, with 150mM NaCl, 0.1mM PMSF, 25% glycerol, 0.1mM EDTA, 0.25mM DTT, 10mM glutathione

Contains

no preservative

Storage conditions

-80°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Dry ice

Product Specific Information

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 µg, 10 µg and 20 µg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 µL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase.

Recombinant full-length human CDK11 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

• Validated in LanthaScreen™ Kinase Binding Assay
• Protocols available for simple use and optimized performance
• Multiple pack sizes available for convenient use

Related Product:
LanthaScreen™ Eu Kinase Binding Assay

Protein Form: Recombinant

The gene accession number for CDK11 (Inactive) is NP_055891.1.

Target Information

CDK11 is a member of the cyclin-dependent kinase 11 family which is known to be essential for eukaryotic cell cycle control and it is in close proximity to CDC2L2, a nearly identical gene in the same chromosomal region. CDK11 is a negative regulator of normal cell cycle progression and Its expression is regulated during murine embryogenesis, and its activity is coordinately regulated with that of p34 (cdc2) during the cell cycle. CDK11 gene may harbor 1 or more tumor suppressor genes that affected by chromosome 1p36 modifications in neuroblastoma. CDK11 could be cleaved by caspases and was demonstrated to play roles in cell apoptosis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CDC-related protein kinase p58; cell division cycle 2-like 1 (PITSLRE proteins); cell division cycle 2-like 2 (PITSLRE proteins); Cell division cycle 2-like protein kinase 1; Cell division cycle 2-like protein kinase 2; Cell division protein kinase 11A; Cell division protein kinase 11B; CLK-1; Cyclin-dependent kinase 11A; Cyclin-dependent kinase 11B; Galactosyltransferase-associated protein kinase p58/GTA; p58 CLK-1; PITSLRE B; PITSLRE protein kinase beta; PITSLRE serine/threonine-protein kinase CDC2L1; PITSLRE serine/threonine-protein kinase CDC2L2

View more View less

Gene Aliases: CDC2L1; CDC2L2; CDC2L3; CDK11; CDK11-p110; CDK11-p46; CDK11-p58; CDK11A; CDK11B; CLK-1; p58; p58CDC2L1; p58CLK-1; p58GTA; PITSLRE; PITSLREA; PITSLREB; PK58

View more View less

UniProt ID: (Human) Q9UQ88, (Human) P21127

View more View less

Entrez Gene ID: (Human) 728642, (Human) 984

View more View less

It has to be done as per old AB suggested Products section.
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0