Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Constitutive Active Receptor, Ligand Binding Domain (CAR-LBD) is a 58.6 kDa recombinant human protein (amino acids 85-348) expressed as a GST fusion protein in baculovirus-infected insect cells. CAR-LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Protein Form: Recombinant, Ligand Binding Domain
The protein encoded by this gene is a type I membrane receptor for group B coxsackieviruses and subgroup C adenoviruses. Pseudogenes of this gene are found on chromosomes 15, 18, and 21.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 46 kD coxsackievirus and adenovirus receptor (CAR) protein; CAR; Coxsackievirus and adenovirus receptor; Coxsackievirus B-adenovirus receptor; CVB3-binding protein; HCVADR
Gene Aliases: CAR; CAR4/6; CXADR; HCAR
UniProt ID: (Human) P78310
Entrez Gene ID: (Human) 1525
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support