Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Proteins & Peptides ›
  • BRCAA1 Proteins

Invitrogen

Human BRCAA1 (aa 659-728) Control Fragment Recombinant Protein

View all (2) BRCAA1 proteins

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Tech Support
Datasheet
Tech Support

Cite Human BRCAA1 (aa 659-728) Control Fragment Recombinant Protein

Product Details

RP-108765

Applications
Tested Dilution
Publications

Control (Ctrl)

Assay-dependent
-

Blocking Assay (BLOCK)

Assay-dependent
-
Product Specifications

Species

Human

Expression System

E. coli

Amino acid sequence

NCKLRRLSKPPFQTNPSPEMVSKLDLTDAKNSDTAHIKSIEITSILNGLQASESSAEDSEQEDERGAQDM

Tag

His-ABP-tag

Class

Recombinant

Type

Protein

Purity

>80% by SDS-PAGE and Coomassie blue staining

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

≥5.0 mg/mL

Purification

purified

Storage buffer

1M urea/PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Product Specific Information

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%).

This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139781. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Target Information

The exact function of C18orf25 remains unknown. There are two named isoforms.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 180 kDa Sin3-associated polypeptide; ARID domain-containing protein 4B; AT rich interactive domain 4B (RBP1-like); AT-rich interactive domain-containing protein 4B; breast cancer-associated antigen 1; Breast cancer-associated antigen BRCAA1; breast carcinoma-associated antigen; Histone deacetylase complex subunit SAP180; Rb-binding protein homolog; Retinoblastoma-binding protein 1-like 1; Sin3-associated polypeptide p180; SIN3A-associated protein 180

View more View less

Gene Aliases: ARID4B; BCAA; BRCAA1; RBBP1L1; RBP1L1; SAP180

View more View less

UniProt ID: (Human) Q4LE39

View more View less

Entrez Gene ID: (Human) 51742

View more View less

It has to be done as per old AB suggested Products section.
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0