Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Proteins & Peptides ›
  • BIVM Proteins

Invitrogen

Human BIVM (aa 9-109) Control Fragment Recombinant Protein

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Tech Support
Datasheet
Tech Support

Cite Human BIVM (aa 9-109) Control Fragment Recombinant Protein

Product Details

RP-97956

Applications
Tested Dilution
Publications

Control (Ctrl)

Assay-dependent
-

Blocking Assay (BLOCK)

Assay-dependent
-
Product Specifications

Species

Human

Expression System

E. coli

Amino acid sequence

RSNDSGNGEHKSERKSPEENLQGAVKSFCTSASGAPLGPKGDGHYPWSCPVTHTREKIYAICSDYAFLNQATSIYKTPNPSRSPCLPDSTSLSAGNNSSRY

Tag

His-ABP-tag

Class

Recombinant

Type

Protein

Purity

>80% by SDS-PAGE and Coomassie blue staining

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

≥5.0 mg/mL

Purification

purified

Storage buffer

1M urea/PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Product Specific Information

Highest antigen sequence indentity to the following orthologs: Mouse (76%), Rat (76%).

This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62140. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Target Information

BIVM has been identified using an electronic search based on the conservation of short sequence motifs within the variable region of immunoglobulin (Ig) genes. It is tightly linked (41 bp) and in the opposite transcriptional orientation to MGC5302 (also known as KDEL1 and EP58) in human. BIVM has two isoforms with MW 57 kDa and 32 kDa.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Basic immunoglobulin-like variable motif-containing protein; RP11-484I6.1

View more View less

Gene Aliases: BIVM

View more View less

UniProt ID: (Human) Q86UB2

View more View less

Entrez Gene ID: (Human) 54841

View more View less

It has to be done as per old AB suggested Products section.
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0