Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Proteins & Peptides ›
  • ANKK1 Proteins

Invitrogen

Human ANKK1, GST Tag Recombinant Protein

View all (2) ANKK1 proteins

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Tech Support
Datasheet
Tech Support

Cite Human ANKK1, GST Tag Recombinant Protein

Product Details

A30970

Applications
Tested Dilution
Publications

Kinase Assay (KA)

Assay-dependent
-
Product Specifications

Species

Human

Expression System

Insect cells

Amino acid sequence

Full length

Tag

GST-tag

Molecular weight

110.9 kDa

Class

Recombinant

Type

Protein

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

purified

Storage buffer

50mM tris HCl, pH 7.5, with 10mM glutathione, 25% glycerol, 0.25mM DTT, 0.1mM PMSF, 150mM NaCl, 0.1mM EDTA

Contains

no preservative

Storage conditions

-80°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Dry ice

Product Specific Information

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 µg, 10 µg and 20 µg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 µL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase.

This ANKK1 recombinant human protein (full length) was expressed in insect cells.

The recombinant full length ANKK1 can be used in a number of different applications, but has been specifically validated for use with our LanthaScreen™ Kinase binding assay technology to identify novel kinase inhibitors. ANKK1 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service.

• Validated for use with the LanthaScreen™ Eu Kinase Binding assay
• Available in multiple pack sizes for convenient use
• Also available in our SelectScreen Kinase Profiling Service

Protein Form: Full Length, Recombinant, Active

The gene accession number for ANKK1 is NP_848605.1.

Target Information

ANKK1 serine/threonine kinase is a member of the RIPK family and is closely linked to the DRD2 gene. ANKK1 is closely linked to DRD2 gene on chr 11, and a well studied restriction fragment length polymorphism designated TaqIA, was originally associated with the DRD2 gene, however, later was determined to be located in exon 8 of ANKK1 gene, where it causes a nonconservative amino acid substitution. It is not clear if ANKK1 plays any role in neuropsychiatric disorders previously associated with Taq1A RFLP.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Ankyrin repeat and protein kinase domain-containing protein 1; Protein kinase PKK2; SgK288; Sugen kinase 288; X-kinase

View more View less

Gene Aliases: ANKK1; PKK2; SGK288

View more View less

UniProt ID: (Human) Q8NFD2

View more View less

Entrez Gene ID: (Human) 255239

View more View less

It has to be done as per old AB suggested Products section.
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0