Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Proteins & Peptides ›
  • ACVR2B Proteins

Invitrogen

Human ACVR2B, GST Tag Recombinant Protein

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Tech Support
Datasheet
Tech Support

Cite Human ACVR2B, GST Tag Recombinant Protein

Product Details

PV6049

Applications
Tested Dilution
Publications

Kinase Assay (KA)

Assay-dependent
-
Product Specifications

Species

Human

Expression System

Baculovirus

Amino acid sequence

185-488

Tag

GST-tag

Molecular weight

62 kDa

Class

Recombinant

Type

Protein

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

purified

Storage buffer

50mM tris

Contains

no preservative

Storage conditions

-80°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Dry ice

Product Specific Information

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 µg, 10 µg and 20 µg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 µL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase.

Protein Form: Catalytic

Target Information

ACVR2B is a type II member of the TGF-beta family of receptor ser/thr kinases and is involved in the activin and myostatin signaling pathway. Activins and inhibins are members of the TGF-beta superfamily due to amino acid homology with respect to the conservation of 7 of the 9 cysteine residues common to all TGF-beta forms. To date, seven type I and five type II activin receptors have been cloned from mammals, including activin receptor IA, activin receptor IIA, activin receptor IB, and activin receptor IIB. In addition, two splice variants of activin receptor IIA and five splice variants of activin receptor IIB have been reported.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: activin A receptor type IIB; activin A receptor, type IIB; Activin receptor type IIB; Activin receptor type-2B; ACTR-IIB; MGC116908

View more View less

Gene Aliases: ActR-IIB; ACTRIIB; ACVR2B; HTX4

View more View less

UniProt ID: (Human) Q13705

View more View less

Entrez Gene ID: (Human) 93

View more View less

It has to be done as per old AB suggested Products section.
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-74848bcbc5-9vj6h:80/100.66.131.58:80.
git-commit: ee75265227445a99bc079a1fecc2a603f2c9394f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0