This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BAIAP2 SIRNA Assays ›
Gene Symbol:
Baiap2
Gene Name:
brain-specific angiogenesis inhibitor 1-associated protein 2
Gene Aliases:
IRSp53, R75030
Chromosome Location:
Chr.11: 119942869 - 120006782 on Build GRCm38
UniGene ID:
Mm.197534
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s99139

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Baiap2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037754.3 NP_001032843.1 7 729
NM_001037755.3 NP_001032844.2 7 729
NM_130862.4 NP_570932.2 7 729
GenBank mRNA AF390178.1 - 7 687
AF390179.1 - 7 687
AK049469.1 - 7 730
AK145924.1 - 7 730
AK158579.1 - 7 754
AK160401.1 - 7 730
AK205197.1 - 1 93
BC006620.1 - 7 720 3158004
BC016411.1 - 7 720 3158004

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

plasma membrane organization
Rho protein signal transduction
insulin receptor signaling pathway
regulation of cell shape
response to bacterium
dendrite development
positive regulation of actin filament polymerization
regulation of actin cytoskeleton organization
regulation of synaptic plasticity
actin filament bundle assembly
actin crosslink formation
positive regulation of dendritic spine morphogenesis
positive regulation of actin cytoskeleton reorganization
protein binding
cytoskeletal adaptor activity
PDZ domain binding
identical protein binding
proline-rich region binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0