This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SFT2D1 SIRNA Assays ›
Gene Symbol:
Sft2d1
Gene Name:
SFT2 domain containing 1
Gene Aliases:
5630401J11Rik, AA409698
Chromosome Location:
Chr.17: 8311103 - 8327442 on Build GRCm38
UniGene ID:
Mm.311080
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s98644

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sft2d1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134114.2 NP_598875.2 4 367
GenBank mRNA AK017471.1 - 2 111
AK151338.1 - 4 298
AK171776.1 - 2 132
BC006777.1 - 4 322
BC091770.1 - 4 368 4974098
BC130232.1 - 4 291 40141936

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_145169, AF041429, BC018969, BC028388.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0