This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ODF2 SIRNA Assays ›
Gene Symbol:
ODF2
Gene Name:
outer dense fiber of sperm tails 2
Gene Aliases:
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location:
Chr.9: 128455155 - 128501292 on Build GRCh38
UniGene ID:
Hs.129055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s9827

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ODF2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242352.1 NP_001229281.1 5 758
NM_001242353.1 NP_001229282.1 5 844
NM_002540.4 NP_002531.3 7 1046
NM_153432.1 NP_702910.1 4 604
NM_153433.1 NP_702911.1 5 771
NM_153435.1 NP_702913.1 5 664
NM_153436.1 NP_702914.1 4 428
NM_153437.2 NP_702915.1 5 694
NM_153439.1 NP_702917.1 4 547
NM_153440.1 NP_702918.1 4 474
XM_005252010.2 XP_005252067.1 4 556
XM_005252011.2 XP_005252068.1 4 496
XM_005252012.3 XP_005252069.1 5 746
XM_005252013.3 XP_005252070.1 5 693
XM_005252014.3 XP_005252071.1 5 742
XM_005252017.4 XP_005252074.1 5 647
XM_005252018.2 XP_005252075.1 6 852
XM_005252019.3 XP_005252076.1 6 871
XM_005252022.2 XP_005252079.1 5 670
XM_005252023.2 XP_005252080.1 5 641
XM_005252024.3 XP_005252081.1 5 660
XM_005252026.4 XP_005252083.1 5 589
XM_005252029.2 XP_005252086.1 5 670
XM_006717126.3 XP_006717189.1 5 746
XM_011518722.2 XP_011517024.1 5 670
XM_011518723.2 XP_011517025.1 5 613
XM_011518724.2 XP_011517026.1 6 834
XM_011518725.2 XP_011517027.1 6 838
XM_011518726.2 XP_011517028.1 6 856
XM_011518728.2 XP_011517030.1 5 607
XM_011518729.2 XP_011517031.1 5 543
XM_011518730.2 XP_011517032.1 5 670
XM_011518732.2 XP_011517034.1 5 456
XM_011518744.1 XP_011517046.1 4 496
XM_017014764.1 XP_016870253.1 6 827
XM_017014765.1 XP_016870254.1 6 846
XM_017014766.1 XP_016870255.1 4 556
XM_017014767.1 XP_016870256.1 6 838
XM_017014768.1 XP_016870257.1 5 630
XM_017014769.1 XP_016870258.1 5 659
XM_017014770.1 XP_016870259.1 6 814
XM_017014771.1 XP_016870260.1 6 795
XM_017014772.1 XP_016870261.1 6 871
XM_017014773.1 XP_016870262.1 6 847
XM_017014774.1 XP_016870263.1 5 557
XM_017014775.1 XP_016870264.1 6 814
XM_017014776.1 XP_016870265.1 6 795
XM_017014777.1 XP_016870266.1 5 597
GenBank mRNA AF012549.1 - 5 634
AF053970.1 - 3 333
AK093367.1 - 4 554
AK299303.1 - 5 606
AK301754.1 - 4 471
AK301842.1 - 4 396
AK301855.1 - 4 450
AK302684.1 - 5 698
AK309917.1 - 5 549
AY319414.1 - 4 580
AY366499.1 - 5 541
BC010629.1 - 7 844 3874501
BC091500.1 - 5 658 30378678
BI831601.1 - 5 685
BM563800.1 - 528
DB044506.1 - 4 531
DC397869.1 - 4 428
DQ444713.1 - 3 371
DQ444714.1 - 3 314

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: DQ091767, DQ091770.

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
multicellular organism development
spermatid development
structural molecule activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0