This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other COG4 SIRNA Assays ›
Gene Symbol:
Cog4
Gene Name:
component of oligomeric golgi complex 4
Gene Aliases:
AW554810, D8Ertd515e
Chromosome Location:
Chr.8: 110846675 - 110882234 on Build GRCm38
UniGene ID:
Mm.272930
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s97932

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cog4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133973.2 NP_598734.1 11, 12 1504
GenBank mRNA AK156466.1 - 11, 12 1496
AK172465.1 - 11, 12 1495
AK179605.1 - 1, 2 88
AK188268.1 - 1, 2 90
AK188304.1 - 1, 2 88
AY223866.1 - 11, 12 1531
BC023120.1 - 11, 12 1481 4161858

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108449.

Gene Ontology Categories:

Function(s) Process(es)

transport
retrograde vesicle-mediated transport, Golgi to ER
Golgi organization
protein transport
Golgi vesicle prefusion complex stabilization
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0