This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRRT4 SIRNA Assays ›
Gene Symbol:
Prrt4
Gene Name:
proline-rich transmembrane protein 4
Gene Aliases:
AW455481, D330027H18Rik
Chromosome Location:
Chr.6: 29169230 - 29179584 on Build GRCm38
UniGene ID:
Mm.22818
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s97743

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prrt4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101443.1 NP_001094913.1 3 898
GenBank mRNA AK052319.1 - 3 900
BC140954.1 - 3 894 9055566

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0