This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBE3C SIRNA Assays ›
Gene Symbol:
Ube3c
Gene Name:
ubiquitin protein ligase E3C
Gene Aliases:
AI853514, mKIAA0010
Chromosome Location:
Chr.5: 29569242 - 29676077 on Build GRCm38
UniGene ID:
Mm.137746
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s97664

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ube3c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133907.3 NP_598668.1 8 1088
GenBank mRNA AK029973.1 - 8 898
AK045211.1 - 8 886
AK046056.1 - 8 1089
AK122187.1 - 8 1023
BC059245.1 - 8 903
BC120731.1 - 8 835 40129654
BC137626.1 - 8 835 8860645

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0