This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDC123 SIRNA Assays ›
Gene Symbol:
Cdc123
Gene Name:
cell division cycle 123
Gene Aliases:
AA959893, G431001I09Rik
Chromosome Location:
Chr.2: 5794294 - 5844968 on Build GRCm38
UniGene ID:
Mm.181490
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s97339

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdc123
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133837.4 NP_598598.1 11 792
GenBank mRNA AK090122.1 - 11 793
AK141463.1 - 11 780
AK159208.1 - 11 813
BC019753.1 - 11 767 5008314
BC022972.1 - 11 781 5368775
BC023821.1 - 11 784 5354711
BC024787.1 - 11 779 5359215

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006023, CR456966, BC001600, D14878, AK225634, and others.

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
cell division
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0