This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC24A3 SIRNA Assays ›
Gene Symbol:
Slc24a3
Gene Name:
solute carrier family 24 (sodium/potassium/calcium exchanger), member 3
Gene Aliases:
NCKX3
Chromosome Location:
Chr.2: 145167712 - 145642166 on Build GRCm38
UniGene ID:
Mm.217171
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s97092

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc24a3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053195.2 NP_444425.1 12 1110
GenBank mRNA AF314821.1 - 12 1150
BC005742.1 - 2 240
BC017615.1 - 12 1110 4914291

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: AY009158.

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
potassium ion transport
sodium ion transport
calcium ion transport
cellular calcium ion homeostasis
sodium ion transmembrane transport
transmembrane transport
calcium ion transmembrane transport
potassium ion transmembrane transport
calcium, potassium:sodium antiporter activity
symporter activity
antiporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0