This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DDX50 SIRNA Assays ›
Gene Symbol:
Ddx50
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 50
Gene Aliases:
4933429B04Rik, 8430408E17Rik, GU2, RH-II
Chromosome Location:
Chr.10: 62616015 - 62651606 on Build GRCm38
UniGene ID:
Mm.114116
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s97030

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ddx50
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053183.2 NP_444413.1 6 857
GenBank mRNA AF334104.1 - 6 858
AK030239.1 - 7 929
AK041281.1 - 6 756
BC049906.1 - 6 858
BC055440.1 - 6 851
BC075714.1 - 6 809
BC090996.1 - 6 877 6757408

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001013198.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
RNA secondary structure unwinding
nucleotide binding
nucleic acid binding
RNA binding
ATP-dependent RNA helicase activity
helicase activity
ATP binding
hydrolase activity
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0